Learn More
Macherey-Nagel™ Silica Chromabond PCA Adsorbent
Shop All Macherey-Nagel Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Quantity | 100 g |
| For Use With (Application) | Strong Cations |
| Product Type | Chromabond PCA Adsorbent |
| Particle Size | 45 μm |
| Material | Silica |
| pH Range | 2 to 8 |
| Pore Size | 60 Å |
| Surface Area (Metric) | 500 m2/g |