Learn More
Trajan Scientific and Medical™ SilFlow™ Replacement Fused Silica Tubing
Microfluidic platform for multi-dimensional GC.
Brand:  Trajan Scientific and Medical™ 123752
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15209193
  • 590.00 NOK / Each
Estimated Shipment: 31-05-2024
Description
  • Enhanced GC connectivity
  • Replacement ferrules and tubing available
Specifications
Specifications
SilFlow Replacement Fused Silica Tubing 0.363mm OD x 0.15mm ID x 2m Length
0.15 mm
2 m
Replacement Fused Silica Tubing
0.363 mm