Learn More
Thermo Scientific™ SilTite™ Replacement Ferrules , Nuts and Baseplate Seals
Product Code. 10608515 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Agilent SSL Injectors
All SilTite Kits
MS Interface
Thermo Scientific DSQ GC-MS Interface
Thermo Scientific DSQ GC-MS Interface Kits
Thermo Scientific TRACE 1300/1310 SSL and ISQ GC-MS Interface Kits
Includes:
Pk. of 10 Ferrules
Pk. of 5 Nuts
Description:
290 MA201
290 MA202
290 MA203
290 MA205
290 MA207
290 MT211
290 MT221
290 MT222
290 MT223
Unit Size:
Pack of 10
Pack of 5
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Includes Description unitSize
10608515 Thermo Scientific TRACE 1300/1310 SSL and ISQ GC-MS Interface Kits Pk. of 10 Ferrules 290 MA201 Pack of 10
10670616 All SilTite Kits Pk. of 10 Ferrules 290 MA202 Pack of 10
10659275 All SilTite Kits Pk. of 10 Ferrules 290 MA203 Pack of 10
10724077 MS Interface Pk. of 5 Nuts 290 MA205 Pack of 10
10391145 Agilent SSL Injectors Pk. of 5 Nuts 290 MA207 Pack of 5
10565625 Thermo Scientific DSQ GC-MS Interface Pk. of 5 Nuts 290 MT211 Pack of 10
10698695 Thermo Scientific DSQ GC-MS Interface Kits Pk. of 10 Ferrules 290 MT221 Pack of 10
10713677 Thermo Scientific DSQ GC-MS Interface Kits Pk. of 10 Ferrules 290 MT222 Pack of 10
10321395 Thermo Scientific DSQ GC-MS Interface Kits Pk. of 10 Ferrules 290 MT223 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10608515 Supplier Thermo Scientific™ Supplier No. 290MA201
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
Eliminate leaks with the Thermo Scientific™ SilTite Ferrule.
Eliminate leaks with the Thermo Scientific™ SilTite Ferrule, which forms a strong, permanent, airtight seal around the capillary column.
  • SilTite metal ferrules and nuts are made from the same material and have the same thermal
  • expansion coefficient
  • Base of the ferrule is flat and forms a perfect seal with the MS interface
  • The ferrule's temperature tolerance is well above the limit of the injector, MS interface or GC oven
  • Unlike other ferrules, do not need re-tightening after installation
TRUSTED_SUSTAINABILITY
Specifications
Type Replacement SilTite Ferrules
Description 290 MA201
For Use With (Equipment) Thermo Scientific TRACE 1300/1310 SSL and ISQ GC-MS Interface Kits
Material SilTite Metal
Includes Pk. of 10 Ferrules
Diameter (Metric) Inside 0.1 to 0.25 mm