Learn More
Thermo Scientific™ SilTite™ Replacement Ferrules , Nuts and Baseplate Seals
Shop All Thermo Scientific ProductsDescription
Eliminate leaks with the Thermo Scientific™ SilTite Ferrule, which forms a strong, permanent, airtight seal around the capillary column.
- SilTite metal ferrules and nuts are made from the same material and have the same thermal
- expansion coefficient
- Base of the ferrule is flat and forms a perfect seal with the MS interface
- The ferrule's temperature tolerance is well above the limit of the injector, MS interface or GC oven
- Unlike other ferrules, do not need re-tightening after installation
Specifications
Specifications
| Type | SilTite Nuts |
| Description | 290 MA207 |
| For Use With (Equipment) | Agilent SSL Injectors |
| Includes | Pk. of 5 Nuts |