Learn More
Heidolph™ Boss Head
Product Code. 10124932 Shop All Heidolph Instruments Products
Change view
Click to view available options
Voltage:
115/230 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
10124932 115/230 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10124932 Supplier Heidolph™ Supplier No. 5703110000
  • ONLINE EXCLUSIVE PRICE
    458.00 NOK
    448.22 NOK / Pack of 1
    Save 9.78 NOK
    2% Off
Estimated Shipment: 10-12-2026
Suitable for stand rods with up to 13.1 mm maximum diameter
Boss head made of anodized aluminum, used to fix retort clamps to stand rods for perfect vertical alignment.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Boss Head
Voltage 115/230 V
Width (Metric) 68
Material Anodized Aluminum
For Use With (Application) Fix retort clamps to stand rods for perfect vertical alignment
Holds Stand rods up to 13.1 mm dia.
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.