Learn More
Ambion™ Silencer™ Select Pre-Designed siRNA
Product Code. 15581084
Click to view available options
Purity:
HPLC
HPLC purified
In Vivo
Standard
Standard purity
Quantity:
1 μmol
1 nmol
10 μmol
20 nmol
20 nmol (Inventoried)
250 nmol
40 nmol
40 nmol (Inventoried)
5 nmol
5 nmol (Inventoried)
Unit Size:
1nmol
5nmol
Each
Product Code. 15581084
Brand: Ambion™ 4444246
Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry.
Silencer Select Predesigned siRNAs offer researchers a powerful and reliable tool for gene silencing applications. Designed with advanced algorithms to ensure high specificity and efficiency, these siRNAs offer a reliable and powerful solution for gene silencing applications. Chemically modified with locked-nucleic acid (LNA) modifications to enhance stability and reduce off-target effects, Silencer Select Predesigned siRNAs deliver consistent and reproducible results across a wide range of experimental conditions.
Key features include:
  • Precise silencing—minimize off-target effects with the combination of algorithm design and LNA chemical modifications
  • Enhanced potency—achieve effective gene silencing with lower siRNA concentrations
  • Guaranteed silencing—two out of two predesigned siRNAs will result in gene knockdown
  • Comprehensive support—access detailed protocols and expert technical support
Optimized design for enhanced specificity
Our internally developed algorithm has been fine-tuned to create siRNAs with sequences that maximize on-target effects while minimizing off-target interactions. This optimized design approach significantly reduces the risk of unintended gene silencing, thereby increasing the reliability and interpretability of experimental results. The specificity of Silencer Select siRNAs allows researchers to confidently attribute observed phenotypes to the silencing of their gene of interest.
Effective silencing at lower concentrations
A key advantage of Silencer Select siRNAs is their ability to achieve effective silencing at lower siRNA concentrations. This not only reduces experimental costs but also minimizes the potential for off-target effects that can occur at higher doses. Maintaining high knockdown efficiency at reduced concentrations highlights the potency and precision of our siRNA design.
Streamlined workflow for rapid results
Utilizing our gene knockdown technology is straightforward and efficient. Simply order an siRNA for the desired gene target, deliver it to the cells, and detect knockdown efficiency and validate the resulting phenotype. Take advantage of our validated siRNA designs for better confidence in your experiments. This streamlined process allows for rapid experimental progress and accelerated discovery in gene function studies.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Room temperature
Description Silencer™ Select siRNA, 1μmol, HPLC Purity, The Silencer Select siRNAs are Classic 21-mers Which Incorporate the Latest Improvements in siRNA Design, Off-target Effect Prediction Algorithms and Chemistry
Product Type Silencer™ Select siRNA
Purity HPLC
Quantity 1 μmol