Learn More
Ambion™ Silencer™ Custom Designed siRNA
Product Code. 15581054
Click to view available options
Purity:
HPLC
Standard
Quantity:
20 nmol
40 nmol
5 nmol
Unit Size:
20nmol
40nmol
5nmol
Each
Product Code. 15581054
Brand: Ambion™ 4399673
Silencer™ custom siRNAs (unmodified 21-bp duplexes) are cost-effective RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types.
  • Cost-effective RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types
  • Use the GeneAssist™ Custom siRNA Builder to design and order a custom siRNA
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at or below -20°C. Do not store in a frost-free freezer. (Dried oligonucleotides are shipped at ambient temperature.) Additional Materials Included: 1.75 mL Nuclease-free Water.
Description GeneAssist - Custom Designed Silencer™ siRNA
For Use With (Application) Knocking down gene expression in order to study protein function in a wide range of cell types
Product Line Silencer
Product Type Custom Designed siRNA
Purity HPLC
Quantity 20 nmol
Shipping Condition Ambient
Temperature (Metric) Upon receipt, store in a non-frost-free freezer at or below -20°C