Learn More
BRAND™ SilberBrand™ ETERNA Type 2 Graduated Pipettes
AR-Glas™ graduated pipettes
Brand: BRAND™ 27006
560.50 NOK valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Additional Details : Weight : 0.27000kg
Description
Specifications
To Deliver | |
B | |
12 | |
2 mL |
DIN EN ISO 835 | |
0.02 mL | |
Black | |
360 mm |