Learn More
BRAND™ SILBERBRAND Compact Automatic Burette Spare Burette Tube
Product Code. 10226092 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10226092 25 mL Each
10625903 10 mL Each
10635903 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10226092 Supplier BRAND™ Supplier No. 23836
SILBERBRAND Spare Burette Tube is suitable for use with a compact automatic burette.
  • Constructed of Borosilicate Glass 3.3
  • Clear glass
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Capacity (Metric) 25 mL
For Use With (Equipment) Compact Automatic Burette
Material Borosilicate Glass (3.3)
Type Buret Tube