Learn More
BRAND™ SILBERBRAND Automatic Burette Spare Burette Tube
Product Code. 10218464 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
Color:
Amber
Clear
Unit Size:
Each
Product Code. Capacity (Metric) Color unitSize
10218464 50 mL Amber Each
10606331 25 mL Clear Each
10552964 25 mL Amber Each
3 options
Product Code. 10218464
Brand: BRAND™ 23724
Only null left
Add to basket
Add to basket
SILBERBRAND Spare Burette Tube is suitable for use with an automatic burette.
  • For use with automatic burette
  • Dr. Scilling pattern
TRUSTED_SUSTAINABILITY
Specifications
Description Spare burette f. aut.burette Schilling 50 ml SILBERBRAND Schellb., AR-GLAS™, amber glass, l. 580 mm
Color Amber
Capacity (Metric) 50 mL
Length (Metric) 580 mm
For Use With (Equipment) Automatic Burette
Material Borosilicate Glass (3.3)
Type Spare Buret