Learn More
SIK1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 51045-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16896315
  • 3828.53 NOK / 150µL
Estimated Shipment: 16-10-2023
Description
The SIK1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SIK1. This antibody recognizes human, mouse antigen. The SIK1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
SIK1
Polyclonal
Unconjugated
150 μL
150094
IgG
Western Blot, Immunohistochemistry, Immunofluorescence
133 μg/mL
Rabbit
Primary
Human, Mouse