Learn More
Uvex™ Signal Men Trousers 7432-Yellow
Product Code. p-7232024 Shop All Uvex Products
Click to view available options
Size:
42
44
46
48
50
52
54
56
58
60
62
64
90
94
98
102
106
110
Unit Size:
Each
Product Code. 17206831
Brand: Uvex™ 8965610
Only null left
Add to basket
Add to basket
All that is essential is that everyone can be seen despite the poor visibility. The high-visibility clothing from the uvex protection flash collection uses bright reflective strips. And of course the cargo trousers and softshell jackets with their optimum fit can also be seen in daylight
  • Material: 67% polyester, 33% cotton, approx 270 gsm
  • CORDURA™ at highly stressed parts
  • Two back and two thigh pockets with several tool pockets, knee pad pockets on the inner side
  • Certified knee protection class 1 in combination with knee pads (article-no. 9868900)
  • Sizes: 42 - 64, 90 - 110
  • Oversize surcharge: from size 56 and size 106 + 10%
  • Care instructions: 30°C washable, chlorinate impossible, tumble dryer: I point, ironing: I point, dry cleaning: P
  • Certified according to: EN ISO 20471 X2 - EN 14404 - Oekotex 100
TRUSTED_SUSTAINABILITY
Specifications
Size 54
Gender Men's
Product Type Hi-vis Pants
Material Polyester
Color Yellow
Seam Type Taped
No. of Pockets 6
Closure Type Buttons; Zipper
Disposable Reusable
Thickness 270 g/m²