Learn More
Tork Signage Holder for Soap and Sanitiser Dispenser
GREENER_CHOICE
Product Code. p-200113564 Shop All Tork Products
Change view
Click to view available options
Color:
Black
White
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
30243801 Black Pack of 6
30243806 White Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30243801 Supplier Tork Supplier No. 511178
Tork Signage Holder for Tork Soap and Sanitizer Dispensers is a communication tool that encourages good hand hygiene. A perfect way to remind users about the importance of hand hygiene, educate on the correct way to wash their hands or communicate other relevant messages.
  • Promote good hand hygiene with on-dispenser signage for staff and guests.
  • Perfect for reminding users about hand hygiene, showing which refill is in use, educating about the correct way to wash hands, or communicating other relevant messages.
  • Designed to fit S4/S5 Tork soap and sanitiser dispensers.
  • Black or white color creates a seamless and professional appearance when paired with the matching dispenser.
TRUSTED_SUSTAINABILITY
Specifications
Alternate Part Number 7322542116562
Material Plastic
Adhesive Type Non-adhesive
Mounting Type Bracket
For Use With (Application) Promotes Hygiene
Shape Rectangular
Product Type Sign Sleeve
Color Black