missing translation for 'onlineSavingsMsg'
Learn More

Siglec-16 Antibody (706045) [FITC], Novus Biologicals™

Product Code. 18697373 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18697373 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18697373 Supplier Novus Biologicals Supplier No. FAB68191F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Siglec-16 Monoclonal antibody specifically detects Siglec-16 in Human samples. It is validated for Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Siglec-16
Applications Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 706045
Conjugate FITC
Dilution Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias Siglec-P16, SIGLECP16
Host Species Mouse
Immunogen Human Siglec-16 peptide, Accession # A6NMB1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 400709
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.