Learn More
RETSCH Sieving Aids Agate Balls
Brand: RETSCH 32.354.0002
2028.25 NOK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.02000kg
Specifications
Agate Balls | |
10 mm | |
Horizontal Sieve Shaker AS 400 control |
|
10/Pk. |