Learn More
Fisherbrand™ Sieve Brush
A double-ended brush for use with Fisherbrand™ Stainless Steel Test Sieves.
157.00 NOK - 414.00 NOK
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
11502763
|
Fisherbrand™
|
20 x 230 mm |
157.00 NOK
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
11512763
|
Fisherbrand™
|
30 x 135 mm |
414.00 NOK
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
Description
- Provides gentle brushing
- Manufactured from either brass or nylon and bristle
- Two different sizes: 30 x 135mm and 20 x 230mm
Specifications
Term Brush | |
Nylon | |
1 Ea. |