Learn More
SHOWA™ Blue Nitrile N-Dex Medical Exam Gloves
Product Code. 12940935
Change view
Click to view available options
Size:
X-Small
Medium
Large
X-Large
Unit Size:
Pack of 100
Product Code. Size unitSize
12940935 X-Large Pack of 100
12930935 Large Pack of 100
12900935 X-Small Pack of 100
12920935 Medium Pack of 100
4 options
Product Code. 12940935
Brand: SHOWA™ BST60054
Only null left
Add to basket
Add to basket
Protect hands with SHOWA™ Blue Nitrile N-Dex Medical Exam Gloves since they provide three times the puncture resistance of natural rubber or vinyl gloves of comparable thickness. .
  • Powder-free
  • Rolled cuff
  • Blue
  • Disposable
Specifications
Color Blue
Disposable Single-use
Material Nitrile
Size X-Large