Learn More
Greiner Bio-One Shorty Sterile Serological Pipets
GREENER_CHOICE
Product Code. 10679492
Change view
Click to view available options
Volume (Metric):
5 mL
10 mL
Graduations:
1/10
2/10
Unit Size:
Pack of 200
Product Code. Volume (Metric) Graduations unitSize
10679492 10 mL 2/10 Pack of 200
10233992 5 mL 1/10 Pack of 200
2 options
Product Code. 10679492
Brand: Greiner Bio-One 607190
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Drip-free pipetting design with high optical clarity and maximum accuracy.
  • High-grade polystyrene
  • Clear, easily legible graduation
  • Colored vertical stripe for ease in reading the volume
  • With synthetic fibrous filter material for protection against
TRUSTED_SUSTAINABILITY
Specifications
Description Shape: Shorty
Material Polystyrene
Sterility Sterile
Graduated Yes
Graduations 2/10
Packaging Type Individually packed in plastic
Volume (Metric) 10 mL
Type Serological Pipet