Learn More
Greiner Bio-One Shorty Sterile Serological Pipets
Click to view available options
Volume (Metric):
5 mL
10 mL
Graduations:
1/10
2/10
Unit Size:
Pack of 200
Description
- High-grade polystyrene
- Clear, easily legible graduation
- Colored vertical stripe for ease in reading the volume
- With synthetic fibrous filter material for protection against
Specifications
Specifications
| Description | Shape: Shorty |
| Material | Polystyrene |
| Sterility | Sterile |
| Graduated | Yes |
| Graduations | 1/10 |
| Packaging Type | Individually packed in plastic |
| Volume (Metric) | 5 mL |
| Type | Serological Pipet |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion