Learn More
Dräger™ Short-Term Detector Tubes - De Tube Hydrogen Sulfide
Click to view available options
Components Measured:
Hydrogen Sulfide
Hydrogen Sulfide 0.2%/a
Hydrogen Sulfide 0.2/b
Hydrogen Sulfide 2%/A
Hydrogen Sulfide 2/a
Hydrogen Sulfide 2/b
Hydrogen Sulfide, Sulfur Dioxide 0.2%/A
Measurement Range:
0.2 to 5ppm
0.2 to 6ppm
0.2 to 7 vol.%
2 to 200ppm
2 to 40 vol.%
2 to 60ppm
Unit Size:
Each
Pack of 10
Description
Note: Extension of measuring range listed is possible for some detector tubes. Call Fisher Customer Service for more information.
Specifications
Specifications
| Components Measured | Hydrogen Sulfide 2%/A |
| Humidity Range | 3 to 20mg/L |
| Measurement Range | 2 to 40 vol.% |
| No. of Strokes | 1 |
| Quantity | 10 |
| Response Time | 1 min. |
| Temperature Range (English) | 32°F to 104°F |
| Temperature Range (Metric) | 0°C to 40°C |