Learn More
Dräger™ Short-Term Detector Tubes - De Tube Hydrogen Sulfide
Product Code. 10473172
Click to view available options
Components Measured:
Hydrogen Sulfide
Hydrogen Sulfide 0.2%/a
Hydrogen Sulfide 0.2/b
Hydrogen Sulfide 2%/A
Hydrogen Sulfide 2/a
Hydrogen Sulfide 2/b
Hydrogen Sulfide, Sulfur Dioxide 0.2%/A
Measurement Range:
0.2 to 5ppm
0.2 to 6ppm
0.2 to 7 vol.%
2 to 200ppm
2 to 40 vol.%
2 to 60ppm
Unit Size:
Each
Pack of 10
Product Code. 10473172
Brand: Dräger™ 8101961
For gas detection
Note: Extension of measuring range listed is possible for some detector tubes. Call Fisher Customer Service for more information.
Specifications
Components Measured Hydrogen Sulfide 2/b
Humidity Range 20mg/L
Measurement Range 2 to 60ppm
No. of Strokes 1
Quantity 10
Response Time 30 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
compliance-icons
Product Identifier
  • drager tubes hydrogen sulphide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.