Learn More
Dräger™ Short-Term Detector Tubes - De Tube Hydrogen Sulfide
For gas detection
Brand:  Dräger™ CH28101
Additional Details : Weight : 0.03000kg
 View more versions of this product
Product Code. 10153671
  • 2870.00 NOK / Pack of 10
Estimated Shipment: 04-06-2024
Description
Note: Extension of measuring range listed is possible for some detector tubes. Call Fisher Customer Service for more information.
Specifications
Specifications
Hydrogen Sulfide 0.2%/a
0.2 to 7 vol.%
10
32°F to 140°F
40mg/L
1+2
2 min.
0°C to 60°C