Learn More
Dräger™ Short-Term Detector Tubes - De Tube Hydrogen Sulfide
Product Code. 10010022
Change view
Click to view available options
Components Measured:
Hydrogen Sulfide
Hydrogen Sulfide 0.2%/a
Hydrogen Sulfide 0.2/b
Hydrogen Sulfide 2%/A
Hydrogen Sulfide 2/a
Hydrogen Sulfide 2/b
Hydrogen Sulfide, Sulfur Dioxide 0.2%/A
Measurement Range:
0.2 to 5ppm
0.2 to 6ppm
0.2 to 7 vol.%
2 to 200ppm
2 to 40 vol.%
2 to 60ppm
Unit Size:
Each
Pack of 10
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Components Measured Measurement Range Unit Size
10010022 Hydrogen Sulfide 0.2/b 0.2 to 6ppm Pack of 10
10473172 Hydrogen Sulfide 2/b 2 to 60ppm Pack of 10
10347892 Hydrogen Sulfide 0.2 to 5ppm Pack of 10
10388421 Hydrogen Sulfide, Sulfur Dioxide 0.2%/A 0.2 to 7 vol.% Each
10153671 Hydrogen Sulfide 0.2%/a 0.2 to 7 vol.% Pack of 10
12951677 Hydrogen Sulfide 2%/A 2 to 40 vol.% Each
10559722 Hydrogen Sulfide 2/a 2 to 200ppm Each
15365442 Hydrogen Sulfide 2/a 2 to 200ppm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10010022 Supplier Dräger™ Supplier No. 8101991
  • ONLINE EXCLUSIVE PRICE
    2396.25 NOK
    1987.53 NOK / Pack of 10
    Save 408.72 NOK
    17% Off
 Restricted Item
Estimated Shipment: 22-12-2026
For gas detection
Note: Extension of measuring range listed is possible for some detector tubes. Call Fisher Customer Service for more information.
Specifications
Components Measured Hydrogen Sulfide 0.2/b
Type Gas Detection Tube
Humidity Range 20mg/L
Measurement Range 0.2 to 6ppm
No. of Strokes 1
Quantity 10
Response Time 55 sec.
Temperature Range (English) 59°F to 104°F
Temperature Range (Metric) 15° to 40°C