Learn More
Hirschmann™ Short Conical Centrifuge Tube
Click to view available options
Capacity (Metric):
10 mL
Unit Size:
Pack of 100
Description
- Short conical form
- Not graduated
Specifications
Specifications
| Capacity (Metric) | 10 mL |
| Length (Metric) | 100 mm |
| Bottom Shape | Conical |
| Diameter (Metric) | 16 mm |