Learn More
Hanna Instruments™ Shockproof Rubber Boot
Product Code. 12963227
Change view
Click to view available options
Color:
Blue
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
12963227 Blue Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12963227 Supplier Hanna Instruments™ Supplier No. HI710024
  • ONLINE EXCLUSIVE PRICE
    848.75 NOK
    488.32 NOK / Pack of 1
    Save 360.43 NOK
    42% Off
Estimated Shipment: 08-01-2027
Specifications
Product Type Shockproof Rubber Boot
Color Blue
Material Rubber
For Use With (Equipment) Designed to protect your instrument and prevent damage due to accidental impact