Learn More
BINDER™ Shipping Pallet
Product Code. p-200104338 Shop All BINDER Products
Change view
Click to view available options
For Use With (Equipment):
KB260, KB470, KB720, KB1060
KBF260, KBF470, KBF720, KBF1060
KBFPRO260, KBFPRO470, KBFPRO720
KBPRO260, KBPRO470, KBPRO720
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
30238221 KB260, KB470, KB720, KB1060 Each
30238285 KBPRO260, KBPRO470, KBPRO720 Each
30238199 KBF260, KBF470, KBF720, KBF1060 Each
30238282 KBFPRO260, KBFPRO470, KBFPRO720 Each
4 options
Product Code. 30238221
Brand: BINDER™ 80122545
Only null left
Add to basket
Add to basket
Shipping pallet with unloading ramp for installing unit without forklift
For
Shipping pallet with unloading ramp for easy unloading of chamber from shipping pallet.
Specifications
For Use With (Equipment) KB260, KB470, KB720, KB1060
Type Platform
Material Fiber