Learn More
BINDER™ Shipping Pallet
Product Code. 30238221 Shop All BINDER Products
Change view
Click to view available options
For Use With (Equipment):
KB260, KB470, KB720, KB1060
KBF260, KBF470, KBF720, KBF1060
KBFPRO260, KBFPRO470, KBFPRO720
KBPRO260, KBPRO470, KBPRO720
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
30238221 KB260, KB470, KB720, KB1060 Each
30238285 KBPRO260, KBPRO470, KBPRO720 Each
30238199 KBF260, KBF470, KBF720, KBF1060 Each
30238282 KBFPRO260, KBFPRO470, KBFPRO720 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30238221 Supplier BINDER™ Supplier No. 80122545
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
Shipping pallet with unloading ramp for installing unit without forklift
For
Shipping pallet with unloading ramp for easy unloading of chamber from shipping pallet.
Specifications
For Use With (Equipment) KB260, KB470, KB720, KB1060
Type Platform
Material Fiber