Learn More
Thermo Scientific™ Shelves for Heratherm™ Ovens and Incubators
Product Code. p-4412777 Shop All Thermo Scientific Products
Change view
Click to view available options
Depth (English):
11.4 in.
12.9 in.
14.8 in.
19.8 in.
20.2 in.
21.06 in.
Width (English):
11.4 in.
12.9 in.
14.8 in.
19.8 in.
20.2 in.
21.06 in.
38.74 in.
For Use With (Equipment):
Adv Prot OMH400/Gen Inc IGS400, 400l
Adv Prot OMH750/Gen Inc IGS750, 750l
Adv Prot/Gen Incubator 400l
Adv Prot/Gen Incubator 750l
Heratherm Adv Prot Incubator 100l
Heratherm Adv Prot Incubator 180l
Heratherm Adv Prot Oven 750I 400V
Heratherm Adv Protocol Incubator 60l
Heratherm Compact Incubator
Heratherm General Incubator 100L
Heratherm General Incubator 180l
Heratherm General Incubator 60l
Unit Size:
Each
Length (English):
12.9 in.
17.2 in.
20.7 in.
Product Code. Depth (English) Width (English) For Use With (Equipment) unitSize Length (English)
11621571 11.4 in. 11.4 in. Heratherm Compact Incubator Each -
10784222 14.8 in. 14.8 in. Heratherm General Incubator 100L Each 12.9 in.
10600601 12.9 in. 12.9 in. Heratherm General Incubator 180l Each 12.9 in.
10841481 14.8 in. 14.8 in. Heratherm General Incubator 60l Each 12.9 in.
10363940 12.9 in. 12.9 in. Heratherm Adv Prot Incubator 100l Each 17.2 in.
10598290 12.9 in. 12.9 in. Heratherm Adv Protocol Incubator 60l Each 12.9 in.
10160060 19.8 in. 19.8 in. Heratherm Adv Prot Incubator 180l Each 17.2 in.
12665316 21.06 in. 21.06 in. Adv Prot/Gen Incubator 400l Each 20.7 in.
12675316 21.06 in. 38.74 in. Adv Prot/Gen Incubator 750l Each -
12685316 21.06 in. 21.06 in. Adv Prot OMH400/Gen Inc IGS400, 400l Each -
12605326 20.2 in. 20.2 in. Heratherm Adv Prot Oven 750I 400V Each -
12695316 21.06 in. 21.06 in. Adv Prot OMH750/Gen Inc IGS750, 750l Each -
12 options
Product Code. 11621571
Brand: Thermo Scientific™ 50125605
Only null left
Add to basket
Add to basket
Perforated and wire mesh shelves
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Heratherm Compact Incubator
Type Shelf
Material Stainless Steel
No. of Shelves 1
Length (Metric) 18.03 cm
Width (English) 11.4 in.
Width (Metric) 18.03 cm
Depth (English) 11.4 in.
Depth (Metric) 28.9 cm