Learn More
- Home
- Products
- Ovens and Furnaces
- Oven Accessories
- Thermo Scientific™ Shelves for Heratherm™ Ovens and Incubators
Thermo Scientific™ Shelves for Heratherm™ Ovens and Incubators
Product Code. 12605326 Shop All Thermo Scientific Products
Change view
Click to view available options
Depth (English):
11.4 in.
12.9 in.
14.8 in.
19.8 in.
20.2 in.
21.06 in.
Width (English):
11.4 in.
12.9 in.
14.8 in.
19.8 in.
20.2 in.
21.06 in.
38.74 in.
For Use With (Equipment):
Adv Prot OMH400/Gen Inc IGS400, 400l
Adv Prot OMH750/Gen Inc IGS750, 750l
Adv Prot/Gen Incubator 400l
Adv Prot/Gen Incubator 750l
Heratherm Adv Prot Incubator 100l
Heratherm Adv Prot Incubator 180l
Heratherm Adv Prot Oven 750I 400V
Heratherm Adv Protocol Incubator 60l
Heratherm Compact Incubator
Heratherm General Incubator 100L
Heratherm General Incubator 180l
Heratherm General Incubator 60l
Unit Size:
Each
Length (English):
12.9 in.
17.2 in.
20.7 in.
12 product options available for selection
| Product Code. | Depth (English) | Width (English) | For Use With (Equipment) | Unit Size | Length (English) |
|---|---|---|---|---|---|
| 12605326 | 20.2 in. | 20.2 in. | Heratherm Adv Prot Oven 750I 400V | Each | - |
| 11621571 | 11.4 in. | 11.4 in. | Heratherm Compact Incubator | Each | - |
| 10784222 | 14.8 in. | 14.8 in. | Heratherm General Incubator 100L | Each | 12.9 in. |
| 10600601 | 12.9 in. | 12.9 in. | Heratherm General Incubator 180l | Each | 12.9 in. |
| 10841481 | 14.8 in. | 14.8 in. | Heratherm General Incubator 60l | Each | 12.9 in. |
| 10363940 | 12.9 in. | 12.9 in. | Heratherm Adv Prot Incubator 100l | Each | 17.2 in. |
| 10598290 | 12.9 in. | 12.9 in. | Heratherm Adv Protocol Incubator 60l | Each | 12.9 in. |
| 10160060 | 19.8 in. | 19.8 in. | Heratherm Adv Prot Incubator 180l | Each | 17.2 in. |
| 12665316 | 21.06 in. | 21.06 in. | Adv Prot/Gen Incubator 400l | Each | 20.7 in. |
| 12675316 | 21.06 in. | 38.74 in. | Adv Prot/Gen Incubator 750l | Each | - |
| 12685316 | 21.06 in. | 21.06 in. | Adv Prot OMH400/Gen Inc IGS400, 400l | Each | - |
| 12695316 | 21.06 in. | 21.06 in. | Adv Prot OMH750/Gen Inc IGS750, 750l | Each | - |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
This item is not returnable.
View return policy
Product Code.
12605326
Supplier
Thermo Scientific™
Supplier No.
50135244
-
3925.00 NOK
3650.02 NOK / Pack of 1
Save 274.98 NOK
7% Off
You must Sign In and have a business account to purchase this item.
Restricted Item
In Stock
Log in to see stock.
Log in to see stock.
Add to basket
3285.02 NOK valid until 2026-10-31
BEST PRICE on Promo! Use promo code "28026" to get your promotional price.
BEST PRICE on Promo! Use promo code "28026" to get your promotional price.
Add to basket
This item is not returnable.
View return policy
Description
Perforated and wire mesh shelves
Specifications
Specifications
| For Use With (Equipment) | Heratherm Adv Prot Oven 750I 400V |
| Type | Shelf |
| Includes | (2) Shelf Supports |
| Width (English) | 20.2 in. |
| Width (Metric) | 51.3 cm |
| Depth (English) | 20.2 in. |
| Depth (Metric) | 51.3 cm |