Learn More
Sheep anti-Human IgA/Secretory Component Secondary Antibody, Novus Biologicals™
Product Code. 18603307 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18603307 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18603307 Supplier Novus Biologicals Supplier No. NBP269294
Sheep Polyclonal Antibody
IgA/Secretory Component Polyclonal antibody specifically detects IgA/Secretory Component in Human samples. It is validated for Immunoelectrophoresis
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgA/Secretory Component
Applications Immunoelectrophoresis
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Formulation Whole antiserum
Host Species Sheep
Immunogen Highly purified IgA/Secretory Component isolated from pooled human milk. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Whole Antiserum
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Reconstitution Reconstitute the lyophilized antiserum by adding 1 mL sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2).
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Lyophilized