Learn More
Sheep anti-Guinea Pig IgM Fc Secondary Antibody, Novus Biologicals™
Product Code. 18160423 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18160423 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18160423 Supplier Novus Biologicals Supplier No. NBP269694
Sheep Polyclonal Antibody
IgM Fc Polyclonal antibody specifically detects IgM Fc in Guinea Pig samples. It is validated for Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgM Fc
Applications Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunoprecipitation 1:10 - 1:500
Formulation Whole antiserum with No Preservative
Host Species Sheep
Immunogen Highly purified normal IgM isolated from pooled guinea pig serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Whole Antiserum
Quantity 1 mL
Primary or Secondary Secondary
Test Specificity The reactivity of the antiserum is restricted to the Fc part of the IgM molecule. In immunoelectrophoresis and radial immunodiffusion, using various antiserum concentrations against normal guinea pig serum a single precipitin line is obtained which shows a reaction of identity with the precipitin line obtained with purified IgM. No precipitation reaction is obtained with purified IgG, IgA, and IgG/Fab fragments.
Reconstitution Reconstitute the lyophilized antiserum by adding 1 ml sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2).
Target Species Guinea Pig
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Lyophilized