Learn More
Sharpsguard™ Cyto 1 Polypropylene Disposal Bin, yellow with purple lid, 1L
Specifications
Specifications
| Capacity (Metric) | 1 L |
| Closure Type | Hinged Lid |
| Color | Yellow |
| Disposable | Single-use |
| Includes | Absorbent pads, hinged lid |
| Material | Polypropylene |