Learn More
Sharpsafe™ Polypropylene Medicinal Sharps Container
Product Code. 12983525
Click to view available options
Capacity (Metric):
2.0 L
4 L
13 L
Unit Size:
Each
Product Code. 12983525
Brand: Sharpsafe™ 41005430
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed for use in public access areas such as A&E and Out-patients. Sharpsafe™ Polypropylene Medicinal Sharps Container allows easy disposal of sharps but provides added security for both the user and the general public. The Protected Access range also discourages the use of the sharps container for non sharps waste, avoiding unnecessary waste disposal costs.
  • Easy disposal of sharps
  • Protected Access range 
  • Polypropylene
Specifications
Capacity (Metric) 4 L
Certifications/Compliance BS 7320, AFNOR NX30-500, UN 3291
Closure Type Lid
Closure Color Yellow
Color Yellow
Model 4100
Quantity 1 Ea.
Includes Closure
For Use With (Application) Designed for use in public access areas such as A&E and out-patients