Learn More
SHANK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55059-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to SHANK1.
Seems to be an adapter protein in the postsynaptic density (PSD) of excitatory synapses that interconnects receptors of the postsynaptic membrane including NMDA-type and metabotropic glutamate receptors via complexes with GKAP/PSD-95 and Homer, respectively, and the actin-based cytoskeleton. May play a role in the structural and functional organization of the dendritic spine and synaptic junction.Specifications
| SHANK1 | |
| Polyclonal | |
| Unconjugated | |
| SHANK1 | |
| SHANK1, SPANK 1, SSTR interacting protein, SSTRIP, synamon | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 243961, 50944, 78957 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.42 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| D3YZU1, Q9WV48, Q9Y566 | |
| SHANK1 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |