Learn More
Epredia™ Shandon™ Tube Occluding Clamp Hemostatic Forceps, 4.5 in. (11.4cm)
Product Code. p-4520633 Shop All Epredia Products
Change view
Click to view available options
Length (English):
7 in.
Unit Size:
Each
Product Code. Length (English) unitSize
12766587 7 in. Each
1 options
Product Code. 12766587
Brand: Epredia™ 3120012
Only null left
Add to basket
Add to basket
Epredia™ Shandon Tube Occluding Clamp Hemostatic Forceps are available in premium and standard grades.
  • Straight; smooth jaws
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards, producing the finest instruments available
  • Guaranteed to be free from defects in material and workmanship
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hemostatic Forceps
Grade Standard
Shape Straight
Length (English) 7 in.
Length (Metric) 17.8 cm