Learn More
Epredia™ Shandon™ Suresorb™ Clinical Pads and Formaldehyde Neutralizer Pads
Product Code. p-4521513 Shop All Epredia Products
Click to view available options
Product Type:
Formaldehyde Neutralizer Pad
Length (English):
10 in.
Width (English):
8 in.
Unit Size:
Pack of 400
Pack of 50
Product Code. 12552736
Brand: Epredia™ 5641
Absorb and lock in liquids with Epredia™ Shandon™ Suresorb™ Clinical Pads and Formaldehyde Neutralizer Pads.
  • Fiber resists tearing, even when saturated
  • Formaldehyde neutralizer pads reduce exposure by chemically bonding with formaldehyde, resulting in the encapsulation of vapors—ideal for grossing areas and autopsy work
  • Clinical pads work as surgical cutting board pads, dissecting pads, autopsy table pads and liners for body trays and cadaver bags
TRUSTED_SUSTAINABILITY
Specifications
Product Type Formaldehyde Neutralizer Pad
Filter Type Formaldehyde
Length (English) 10 in.
Quantity 400/Cs.
Width (English) 8 in.
Width (Metric) 203.2 mm