Learn More
Epredia™ Shandon™ Straight Blunt/Sharp Dissecting Scissors, premium, 6.5 in. (16.5cm)
Product Code. p-4520898 Shop All Epredia Products
Click to view available options
Length (Metric):
16.5 cm
Grade:
Standard
Unit Size:
Each
Product Code. 15202290
Brand: Epredia™ 28253
Only null left
Add to basket
Add to basket
Epredia™ Shandon Straight Blunt/Sharp Dissecting Scissors are available in Premium and Standard grades.
Consistent Quality
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards
  • Guaranteed to be free from defects in material and workmanship
TRUSTED_SUSTAINABILITY
Specifications
Product Type Dissecting Scissors
Blade Style Straight
Grade Standard
Length (English) 6.5 in.
Length (Metric) 16.5 cm
Tip Style Blunt/Sharp