Learn More
Epredia™ Shandon™ Stainless-Steel Instrument Trays, 8.9L x 5W x 2 in. D (22.5 x 12.7 x 5.1cm)
Product Code. p-4520607
Click to view available options
Length (English):
8.9 in.
Unit Size:
Each
Product Code. 15951437
Brand: Epredia™ 73092
This item requires a price update prior to being ordered.
Request a Price for this item here.
Epredia™ Shandon Stainless-Steel Instrument Trays are available in two sizes.
  • Constructed of surgical grade stainless steel
Specifications
Product Type Instrument Trays
Material Stainless Steel
Dimensions (L x W x H) 8.9 x 5 x 2 in. (22.3 x 12.7 x 5.1 cm)
Length (English) 8.9 in.
Length (Metric) 22.5 cm
Width (English) 5 in.
Width (Metric) 12.7 cm
Height (English) 2 in.
Height (Metric) 5.1 cm