Learn More
Epredia™ Stainless-Steel Embedding Base Molds
Product Code. p-4524717 Shop All Epredia Products
Change view
Click to view available options
Dimensions (L x W x H):
10 x 10 x 5 mm
24 x 24 x 5 mm
24 x 37 x 9 mm
Unit Size:
Pack of 10
Product Code. Dimensions (L x W x H) unitSize
12687906 10 x 10 x 5 mm Pack of 10
11306820 24 x 24 x 5 mm Pack of 10
12607916 24 x 37 x 9 mm Pack of 10
3 options
Product Code. 12687906
Brand: Epredia™ 6401015
Optimize tissue embedding with the superior thermal exchange of our high-quality, durable Stainless-Steel Embedding Base Molds.
  • Sizes fit most cassettes
  • Superior thermal exchange
  • Rounded corners
TRUSTED_SUSTAINABILITY
Specifications
Product Type Base Mold
Material Stainless Steel
Dimensions (L x W x H) 10 x 10 x 5 mm