Learn More
Epredia™ Peel-A-Way™ Disposable Embedding Molds
Product Code. 12677736 Shop All Epredia Products
Change view
Click to view available options
Dimensions (L x W x H):
22 x 30 x 20 mm
Shape:
Rectangular
Unit Size:
Pack of 288
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Shape Unit Size
12677736 22 x 30 x 20 mm Rectangular Pack of 288
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12677736 Supplier Epredia™ Supplier No. 1830
  • ONLINE EXCLUSIVE PRICE
    5038.79 NOK
    2127.80 NOK / Pack of 288
    Save 2910.99 NOK
    57% Off
 Restricted Item
Estimated Shipment: 05-11-2026
Promote rapid paraffin hardening with Epredia Peel-A-Way Disposable Embedding Molds.
  • Constructed of rigid transparent plastic.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Embedding Mold
Material Plastic
Dimensions (L x W x H) 22 x 30 x 20 mm
Disposable Single-use
Shape Rectangular