Learn More
Epredia™ Shandon™ OG-6 Stain
Product Code. p-4530124 Shop All Epredia Products
Change view
Click to view available options
Quantity:
4L
Unit Size:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10239788 4L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10239788 Supplier Epredia™ Supplier No. 6766006
Obtain maximum performance and maximum value with Epredia™ Shandon™ OG-6 Stain, a biological stain commission-certified dye.
TRUSTED_SUSTAINABILITY
Specifications
Type OG-6 Papanicolaou Stain
Quantity 4L
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P262-Do not get in eyes, on skin, or on clothing.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : methyl alcohol, ethylene glycol, orange-g certified, phosphotungstic acid