Learn More
Epredia™ Shandon™ Octospot™ Accessories
Product Code. 12590047 Shop All Epredia Products
Click to view available options
Type:
Holding Block
Solvent-Resistant Slides, 8 spots
Thick Filter Cards
Unit Size:
Each
Pack of 100
Product Code. 12590047
Brand: Epredia™ 1001641
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
These Epredia™ accessories are for use with Shandon Octospot Starter Packs.
TRUSTED_SUSTAINABILITY
Specifications
Type Thick Filter Cards
For Use With (Equipment) Shandon Octospot Starter Packs