Learn More
Epredia™ Glyo-Fixx™
Product Code. 10602689 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 gal.
5 gal.
125 x 120mL
Product Type:
Fixative
Unit Size:
3.8 L
Each
Pack of 125
Product Code. Quantity Product Type unitSize
10602689 1 gal. Fixative 3.8 L
10249908 5 gal. Fixative Each
12520067 125 x 120mL Fixative Pack of 125
3 options
Product Code. 10602689
Brand: Epredia™ 6764262
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Obtain superior results to neutral buffered formalin with Glyo-Fixx™, a noncarcinogenic, nonsensitizing, and environmentally friendly substitute.
  • Produces distinct and crisp cellular morphology
  • Fixes most antigens without impairing immunoreactivity
  • Allows special stain dyes, including silver procedures, to be sharp without background staining
  • Presents fine detail in specimen structure
TRUSTED_SUSTAINABILITY
Specifications
Product Type Fixative
Quantity 1 gal.
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Germ cell mutagenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H350-May cause cancer.
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
Supplemental information
  • MIXTURE LIST-Contains : ethyl alcohol, glyoxal, isopropyl alcohol, methyl alcohol