Learn More
Epredia™ Shandon™ Flexible Probe with Eyelet, 6 in. (15.2cm)
Product Code. 15254427 Shop All Epredia Products
Change view
Click to view available options
Length (English):
6 in.
Unit Size:
Each
Product Code. Length (English) unitSize
15254427 6 in. Each
1 options
Product Code. 15254427
Brand: Epredia™ 1101
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Properly hardened, tempered and passivated, this stainless-steel flexible probe is guaranteed to be free from defects in material and workmanship.
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards, producing the finest instruments available
TRUSTED_SUSTAINABILITY
Specifications
Length (English) 6 in.