Learn More
Epredia™ Cryomatrix™ embedding resin
Product Code. 12542716 Shop All Epredia Products
Change view
Click to view available options
Capacity:
120 mL
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
12542716 120 mL Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12542716 Supplier Epredia™ Supplier No. 6769006
  • ONLINE EXCLUSIVE PRICE
    485.00 NOK
    364.75 NOK / Pack of 4
    Save 120.25 NOK
    24% Off
 Restricted Item
Estimated Shipment: 19-11-2026
Secure and support tissue with Epredia™ Cryomatrix™ embedding resin, an outstanding product used to support tissue for frozen sectioning.
  • Forms a firm bond with the specimen holder
  • Freezes quickly, sections smoothly
  • Dissolves in water without residue
TRUSTED_SUSTAINABILITY
Specifications
Capacity 120 mL
Product Type Embedding Media