Learn More
Buerkle™ Shallow Metal Laboratory Bowl
Product Code. p-7224419 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
1 L
2 L
2.5 L
3 L
5 L
6 L
Unit Size:
Each
Product Code. 17564286
Brand: Buerkle™ 42080020
Only null left
Add to basket
Add to basket
This stainless steel bowl is ideal for disinfecting instruments as well as for carrying different devices or samples.
  • For cleaning, disinfecting, sterilizing, or storing laboratory cutlery
  • Stable and hygienic
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1 L
Dimensions (L x W x H) 200 x 150 x 50 mm
Height (Metric) 50 mm
Length (Metric) 200 mm
Material Metal
Width (Metric) 150 mm
Length (English) 7.9 in.
Height (English) 2 in.
Width (English) 5.9 in.
Capacity (English) 0.3 gal.