Learn More
Buerkle™ Shallow Metal Laboratory Bowl
Product Code. 17594286 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
1 L
2 L
2.5 L
3 L
5 L
6 L
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
17594286 3 L Each
17564286 1 L Each
17574286 2 L Each
17584286 2.5 L Each
17504296 5 L Each
17514296 6 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17594286 Supplier Buerkle™ Supplier No. 42080023
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This stainless steel bowl is ideal for disinfecting instruments as well as for carrying different devices or samples.
  • For cleaning, disinfecting, sterilizing, or storing laboratory cutlery
  • Stable and hygienic
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 3 L
Dimensions (L x W x H) 300 x 250 x 50 mm
Height (Metric) 50 mm
Length (Metric) 300 mm
Material Metal
Width (Metric) 250 mm
Length (English) 11.8 in.
Height (English) 2 in.
Width (English) 9.8 in.
Capacity (English) 0.8 gal.