Learn More
Heidolph™ Shaker Vibramax 110
Product Code. 10528951 Shop All Heidolph Instruments Products
Change view
Click to view available options
Speed Range:
150 to 1350 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Range Unit Size
10528951 150 to 1350 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10528951 Supplier Heidolph™ Supplier No. 5443120000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Load Bearing Capacity (Metric) 2 kg
Speed Range 150 to 1350 rpm
Electrical Requirements 230 V
Voltage 230 V
Product Type Shaker
Model Vibramax 110