Learn More
Heidolph™ Rotamax 120 Orbital Shaker
Product Code. 10304491
Change view
Click to view available options
Speed Range:
20 to 300 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Range Unit Size
10304491 20 to 300 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10304491 Supplier Heidolph™ Supplier No. 5444120000
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Performs a gentle motion for sensitive samples and supports culture plates and Erlenmeyer flasks. The Rotamax 120 Orbital Shaker is a compact and space-saving unit with a shaking orbit of 20 mm.
Compact and space-saving unit that accepts load capacities of 2 kg. The shaking orbit of 20 mm performs a gentle motion for sensitive samples and supports culture plates and Erlenmeyer flasks
  • Set and continuously adjust the variable speed on the analog control knob from 20 to 300 rpm
  • An analog process timer allows for unattended operation and can be set from 1 to 120 minutes. When the set time has elapsed an alarm will sound and the operation stops
TRUSTED_SUSTAINABILITY
Specifications
Orbit 20 mm
Load Bearing Capacity (Metric) 2 kg
Speed Range 20 to 300 rpm
Electrical Requirements 230 V
Wattage 33 W
Product Type Shaker
Load Bearing Capacity (English) 4.4 lb.
Certifications/Compliance DIN EN 60529 IP 30
Dimensions (LxW) Platform 220 x 220 mm
Dimensions (L x W x H) 310 x 245 x 125 mm
Weight (English) 12.1 lb.
Weight (Metric) 5.5 kg
Controller Type Electronic
Model Rotamax 120
For Use With (Equipment) Culture Plates, Erlenmeyer Flasks
Includes Non-skid rubber mat
Platform Style Orbital
Timer Range 1 to 120 min.