Learn More
Cole-Parmer™ SH-200 Series See-Saw Rocking Shakers
Product Code. 17721288 Shop All Cole-Parmer Products
Change view
Click to view available options
Load Bearing Capacity (English):
6.6 lb.
22 lb.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Load Bearing Capacity (English) Unit Size
17721288 22 lb. Each
17761288 6.6 lb. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17721288 Supplier Cole-Parmer™ Supplier No. 5190106
  • ONLINE EXCLUSIVE PRICE
    14550.00 NOK
    13784.99 NOK / Pack of 1
    Save 765.01 NOK
    5% Off
View all promotions
Estimated Shipment: 30-11-2026
Gently mix live cultures.
  • Nonslip mat and platform lip hold vessels securely in place
  • Built-in timer and digital speed LED display provide reproducible results
  • Can be used in incubators
These see-saw rocking shakers provide a gentle rocking action at a 7° tilt angle, perfect for culture flasks and Petri dishes. Easily set and view the speed and the timer on the LED display. Control speed from 5 to 70 rpm. The microprocessor maintains speed accuracy even over long runs. Easily set digital timer from 1 to 999 minutes. When timer reaches zero, the shaker automatically stops and sounds an alert.Durable shakers are rated for continuous use. Use in incubators and environmental or cold chambers up to 104°F (40°C).
Specifications
Rocking Angle 7°
Load Bearing Capacity (Metric) 10 kg
Speed Range 5 to 70 rpm
Voltage 230 VAC
Load Bearing Capacity (English) 22 lb.
Frequency 50 Hz
Depth (English) 17.716 in.
Depth (Metric) 45 cm
Height (English) 6.5 in.
Height (Metric) 16.5 cm
Length (English) Platform 14 in.
Length (Metric) Platform 35.5 cm
Width (English) 15.35 in.
Width (English) Platform 14 in.
Width (Metric) 39 cm
Width (Metric) Platform 35.5 cm
Description Large See-Saw Rocker
Display LED
Includes Flat Platform, Nonslip Mat, and Power Cord
Timer Range 1 to 999 minutes
Warranty and Service Offering 3 Year Warranty